Would you like to log in again?
Quadruple Geological Thin Section Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75941-19H 75941-19
102098-296EA 468.77 USD InStock
102098-296 102681-260
Quadruple Geological Thin Section Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
The thin section holder made from aluminum with copper clips and brass screws.
The unit measures 107 x 58 x8 mm and it holds four standard geological thin sections or slides of 47 x 27 mm. The holder has convenient tweezer inserts which allow for easy loading. Clips are provided for secure holding.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty