Would you like to log in again?
Multi Pin Holder for 12 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75952-12 75952-12H
102098-310EA 191.84 USD InStock
102098-310 102098-308
Multi Pin Holder for 12 Pin Stubs, Electron Microscopy Sciences
Specimen Mounting Supplies
This 62 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws.
It accommodates up to twelve standard 12.7 mm pin stubs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty