Would you like to log in again?
45° Multi Holder for 6 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75952-70 75952-70H
102098-326EA 188.58 USD InStock
102098-326 102098-324
45° Multi Holder for 6 Pin Stubs, Electron Microscopy Sciences
Specimen Mounting Supplies
This 35 mm in diameter 45 Degree multi pin holder is designed to save time. It allows for a higher SE signal without having to tilt the specimen It is made from machined aluminum with stainless steel Allen set screws. It accommodates up to six standard 12.7 mm (1/2") pin stubs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty