Would you like to log in again?
Multi Holder for 4 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75953-02J
102098-328EA 105.56 USD InStock
102098-328
Multi Holder for 4 Pin Stubs, Electron Microscopy Sciences
Specimen Mounting Supplies
This multi pin stub adapter for JEOL 32 x 11 mm is designed to save time. It is made from vacuum aluminum with stainless steel set screws.
It accommodates up to fourt standard 12.7 mm pin stubs with 3.2 mm pin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty