Would you like to log in again?
Forensic Certified Mount, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76475-54 76475-50 76475-10 76475-05 76475-53 76475-52
102098-414PK 288.34 USD InStock
102098-414 103302-512 103306-384 102098-412 102098-410 103301-898
Forensic Certified Mount, Electron Microscopy Sciences
Specimen Mounting Supplies
Gun Shot residue mount is free from objectional elements that would interfere with results.
  • Made from special certified aluminum alloy
  • Mounts are 12.7 x 11 mm
  • Available with 8 or 6 mm pin
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty