Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SEM Single Mount Storage Tube and Mailer, Hitachi, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76531-10 76531-05 76535-01
102098-420PK 22.78 USD InStock
102098-420 102098-418 102098-416
SEM Single Mount Storage Tube and Mailer, Hitachi, Electron Microscopy Sciences
Mailers and Shippers
Ideal for storage as well as mailing.
A 15 mm diamter plastic tube and plug is for M4 threaded Hitachi mounts. A 15 mm Hitachi mount can be inserted securely to the plug and then protected by the tube. The internal diameter of the tube is 21 mm (0.83”). The Outer measurements of the tube are: 23 x 52 mm (0.9 x 2.05").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty