Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77010-Y3 77010-R1 77009-G2 77009-W1 77010-W2 77010-O2 77010-G3 77010-Y1 77010-W3 77009-Y2 77010-O3 77010-B2 77009-G3 77009-W2 77010-W1 77010-G2 77011-W3 77009-Y3 77011-W2 77009-G1 77009-Y1 77011-W1 77009-W3 77010-O1 77010-B1 77010-R2 77010-R3 77010-B3 77010-G1 77010-Y2
100488-124RL 25.42 USD InStock
100488-124 100488-146 102098-448 102098-426 100488-148 102098-446 100488-126 102098-444 100488-128 102098-442 100488-140 100488-142 102098-428 100488-122 100488-144 102098-440 100488-156 102098-436 100488-134 100488-136 102098-434 102098-432 100488-138 102098-430 100488-150 100488-152 100488-130 100488-132 102098-438 100488-154
Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
Tapes
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250°F). It may be used as well down to -73°C.
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core. Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty