Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Direct Thermal Cryotags, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Cryo-Tags®
77577-8 77577-7 77577-5 77577-6 77577-3 77577-16 77577-20 77577-2 77577-19 77577-4 77577-18 77577-1 77577-17
102098-546PK 91.88 USD InStock
102098-546 102098-524 102098-534 102098-544 102098-522 102098-520 102098-530 102681-350 102681-352 102098-528 102098-506 102098-538 102098-526
Direct Thermal Cryotags, Electron Microscopy Sciences
Labels
Labels with expanded temperature range and freezable in liquid and vapor phase nitrogen. They adhere to most plastics, glass and metals without cracking, peeling or degrading.
  • Compatible and Economical
  • Temperature resistant from –196 to 70 °C
  • Resistant to scuffing and smearing
Labels accept printing from direct thermal printers such as Dymo LabelWriters. Print popular barcode formats for fast and easy sample identification. Printer setup and use is easy and user friendly. Limit label waste by printing only the quantity required. Labels are now oriented vertically and are compatible with the new LabelWriter 450.
Tubes labeled with Direct Thermal CryoTags easily slide in and out of centrifuge rotors without binding. Users of Direct Thermal Tough-Tags will be happy to know we’ve expanded the temperature range of Tough-Tags and made them Cryo-Tags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty