Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Tough-Spots®
77587-G 77587-W 77585-G 77587-O 77585-O 77588-75 77587-05 77587-B 77587-R 77585-44 77585-R 77585-B 77588-100 77585-Y 77585-W 77585-05 77587-Y
101413-972PK 119.74 USD InStock
101413-972 101413-980 101413-970 101413-976 101413-974 102098-624 101413-968 102098-622 101413-978 102098-620 102098-630 102098-618 102098-628 102098-616 102098-604 102098-626 102098-614
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Labels
Tough-Spots® Labels are pre-cut peel-off labels specially for microcentrifuge tube tops. This chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • Polyolefin labels resist most organic solvents and caustic agents
  • They adhere to most plastics, glass and metals without cracking, peeling, or degrading
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Label sheets are top-coated to readily accept laser printing
  • Assortment includes four sheets of blue, green, red, yellow and orange color
Made of polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty