Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77819-65 77819-25 77819-12
102098-802EA 54.19 USD InStock
102098-802 102098-798 102098-800
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Tapes
EMS introduces this unique double sided carbon tape which is 260 microns in thickness
The base which is polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty