Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Grip Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78144-5 78144-O 78144-3 78144-5A 78144-2A 78144-SS 78144-OO 78144-1 78144-3C 78144-7
102098-834EA 33.11 USD InStock
102098-834 102098-824 102098-826 102098-816 102098-828 102098-818 102098-830 102098-820 102098-832 102098-822
Grip Tweezers, Electron Microscopy Sciences
Forceps
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade.
Style SS measures 135 mm (5 1/3”) Long. M = 8 mm (1/3”) N = 1.5 mm (1/16”)    P = 82 mm (3 ¼”). Style 1 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P =  50 mm (2”). Style 3 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 50 mm (2”). Style 3C measures 110 mm (4 1/3”) Long. M = 9 mm (1/3”) N = 1.5 mm (1/16”)   P = 43 mm (1 2/3”). Style O measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 51 mm (2”). Style OO measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 2.5 mm (1/10”) P = 46 mm (1 ¾”) U = 0.5 mm (½0”).  Style 2A measures 120 mm ( ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 35 mm (1 3/8”) U = 2 mm (1/12”).  Style 5 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 30 mm (1 1/16”).  Style 5A measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).  Style 7 measures 120 mm (4 ¾”) Long. M = 11 mm (½”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty