Would you like to log in again?
Ion Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78146-5 78146-2A 78146-3 78146-OO 78146-1 78146-7 78146-SS 78146-3C 78146-O 78146-5A
102098-846EA 50.82 USD InStock
102098-846 102098-836 102098-848 102098-838 102098-850 102098-840 102098-852 102098-842 102098-854 102098-844
Ion Tweezers, Electron Microscopy Sciences
Forceps
These tweezers feature a special surface coating which releases silver ions. These ions act as an anti-bacterial agent which helps the user from contamination of any pathogens.
Unlike other methods, the use of ion technology does not give rise to any resistance. Consequently, it represents a risk-free method of guaranteeing hygiene, even at workstations where instruments are used by several different people. Made from finest stainless steel grade, these tweezers are perfect companions for your daily use in the laboratory. (We recommend that safety gloves still be used whenever handling any infectious materials).
Style SS measures 135 mm (5 1/3”) Long. M = 8 mm (1/3”) N = 1.5 mm (1/16”)    P = 82 mm (3 ¼”). Style 1 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P =  50 mm (2”). Style 3 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 50 mm (2”). Style 3C measures 110 mm (4 1/3”) Long. M = 9 mm (1/3”) N = 1.5 mm (1/16”)   P = 43 mm (1 2/3”). Style O measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 51 mm (2”).  Style OO measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 2.5 mm (1/10”) P = 46 mm (1 ¾”) U = 0.5 mm (½0”).  Style 2A measures 120 mm ( ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 35 mm (1 3/8”) U = 2 mm (1/12”).  Style 5 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 30 mm (1 1/16”). Style 5A measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).  Style 7 measures 120 mm (4 ¾”) Long. M = 11 mm (½”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty