Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Axal™ Tweezers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
78147-4 78147-7 78147-SS 78147-1 78147-0C11 78147-7A 78147-2A 78147-F 78147-3C 78147-3 78147-5 78147-2 78147-0 78147-5A 78147-4AB 78147-0C
102098-880EA 69.47 USD InStock
102098-880 102098-856 102098-878 102098-868 102098-858 102098-870 102098-860 102098-882 102098-872 102098-862 102098-884 102098-874 102098-886 102098-864 102098-876 102098-866
Axal™ Tweezers, Electron Microscopy Sciences
Forceps
Axal has high yield strength and is 100% antimagnetic. It is highly resistant to corrosion, stress corrosion cracking and hydrogen embattlement.

Style SS: 135 mm Long. M = 8 mm N = 1.5 mm P = 82 mm
Style 1: 120 mm Long. M = 10 mm N = 1.5 mm P = 50 mm
Style 3: 120 mm Long. M = 10 mm N = 1.5 mm P = 50 mm
Style 3C: 110 mm Long. M = 9 mm N = 1.5 mm P = 43 mm
Style 0: 120 mm Long. M = 10 mm N = 1.5 mm P = 51 mm
Style 0C 11: 110 mm Long. M = 9 mm N = 1.5 mm P = 47 mm
Style 4AB: 110 mm Long. M = 10 mm N = 1.5 mm P = 30 mm
Style F: 120 mm Long. M = 10 mm N = 1.5 mm P = 33 mm
Style 2A: 120 mm Long. M = 10 mm N = 1.5 mm P = 35 mm U = 2 mm
Style 2: 115 mm Long. M = 10 mm , N = 1.5 mm P = 34 mm
Style 5: 120 mm Long. M = 10 mm N = 1.5 mm P = 30 mm
Style 4: 110 mm Long. M = 10 mm N = 1.5 mm P = 30 mm
Style 5A: 115 mm Long. M = 10 mm N = 1.5 mm P = 34 mm
Style 7: 120 mm Long. M = 11 mmN = 1.5 mm P = 34 mm
Style 7A: 115 mm Long. M = 10 mm N = 1.5 mm P = 34 mm

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty