Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nano Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78148-5 78148-3C 78148-5A 78148-0 78148-00 78148-SS 78148-3 78148-1 78148-7 78148-2A
102098-900EA 59.85 USD InStock
102098-900 102098-902 102098-904 102098-890 102098-906 102098-892 102098-894 102098-896 102098-898 102098-888
Nano Tweezers, Electron Microscopy Sciences
Forceps
Ultra-modern nanotechnology closes even the tiniest of pores on the surface and this innovation has resulted in the cleanest tweezers surface ever produced. Made from the  finest stainless steel grade, the Lotus effect repels dirt, oil, water and other liquids. These tweezers are a perfect companion in the clean room or any sensitive laboratory areas where contaminant free tools is required. Style SS is 135 mm (5 1/3”) Long. M = 8 mm (1/3”) N = 1.5 mm (1/16”)    P = 82 mm (3 ¼”). Style 1 is 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P =  50 mm (2”). Style 3 is 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 50 mm (2”). Style 3C is 110 mm (4 1/3”) Long. M = 9 mm (1/3”) N = 1.5 mm (1/16”)   P = 43 mm (1 2/3”). Style 0 is 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 51 mm (2”).  Style 00 is 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 2.5 mm (1/10”) P = 46 mm (1 ¾”) U = 0.5 mm (½0”). Style 2A is 120 mm ( ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 35 mm (1 3/8”) U = 2 mm (1/12”). Style 5 is 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 30 mm (1 1/16”). Style 5A is 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).  Style 7 is 120 mm (4 ¾”) Long. M = 11 mm (½”) N = 1.5 mm (1/16”)   P = 34 mm (1 1/3”).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty