Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Swiss Tweezer; Precision, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78170-1G 78170-2G 78170-4G 78170-2AG 78170-5AG 78170-3CG 78170-3G 78170-5ARG 78170-7G 78170-5G
102098-944EA 43.41 USD InStock
102098-944 102098-956 102098-946 102098-958 102098-948 102098-950 102098-940 102098-952 102098-942 102098-954
Swiss Tweezer; Precision, Electron Microscopy Sciences
Forceps
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
  • Tweezers engineered for a NON-SLIP GRIP
  • Light weight –prevents hand fatigue
  • Sensuous touch and functional design
  • Ultra-fine precision tips
  • Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty