Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubis Specialty Tweezers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
78174-6151 78174-B00 78180-1K6 78174-615 78174-524 78174-LB 78174-232 78174-616
102098-966EA 36.32 USD InStock
102098-966 102098-968 102098-980 102098-970 102098-960 102098-972 102098-962 102098-964
Rubis Specialty Tweezers, Electron Microscopy Sciences
Forceps
These tweezers are long and strong.

  • Knurled Handle – non slip
  • Serrated tips and with or without guide pin

Model 232, SA: 158 mm (6 2/9") long. M= 9 mm (1/3"), N = 2 mm (1/12"), P= 80 mm (3 1/8"), R = 58 mm (2 2/3"), S = 44 mm (1 7/9"), T = 17 mm (2/3"), U = 1.7 mm (1/16")
Model 524, SA: 137 mm (5 1/3") long. M= 10 mm (1/3"), N = 1.5 mm (1/16"), P= 60 mm (2 1/3"), R = 52 mm (2"), S = 35 mm (1 3/8"), T = 15 mm (2/3"), U = 1 mm (1/25")
Model 615, SA: 155 mm (6") long. M= 8 mm (1/3"), N = 2 mm (1/12"), R = 57 mm (2 1/4"), S = 41 mm (1 2/3"), T = 15 mm (2/3"), U = 1 mm (1/25")
Model 616, SA: 162 mm (6 3/8") long. M= 9 mm (1/3"), N = 2 mm (1/12"), R = 66 mm (2 2/3"), S = 45 mm (1 3/4"), T = 15 mm (2/3"), U = 1 mm (1/25")
Model B00, SA: 140 mm (5 1/2") long. M= 9 mm (1/3"), N = 1.5 mm (1/16"), P= 60 mm (2 1/3"), R = 52 mm (2), S = 35 mm (1 1/3"), T = 17 mm (2/3"), U = 2.5 mm (1/10")
Model L/Black, SA: 158 mm (6 1/4") long. M= 10 mm (1/3"), N = 1.5 mm (1/16"), P= 65 mm (2 1/2"), R = 45 mm (3 1/4"), S = 35 mm (1 3/8"), T = 13 mm (1/2"), U = 1 mm (1/25")
Model 6151, SA: 140 mm (5 1/2") long, with plastic insulated handle. M= 11 mm (1/2"), N = 1.5 mm (1/16"), T = 15 mm (2/3"), U = 2 mm (1/12")

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty