Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ilford Ilfospeed RC Deluxe Paper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83033-25 83032-10 83032-25 83033-10
102099-378PK 547.51 USD InStock
102099-378 102099-376 102099-382 102099-380
Ilford Ilfospeed RC Deluxe Paper, Electron Microscopy Sciences
Film Processing Supplies
This is a direct replacement for Kodak Kodabrome II RC B&W Papers. Premium Quality Graded Paper on a Resin Coated Base. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m2) with a resin coated base. It is suitable for a wide range of applications including commercial, press, industrial, advertising, and display work. Can be used with virtually all safelights for B&W papers. Neutral Tone Graded RC, can be either dish/tray processed or machine processed. Latent image ability.
  • Can be used with virtually all safelights for B&W papers
  • Neutral Tone Graded RC
  • Can be either Dish/Tray Processed or Machine Processed
  • Latent Image Ability
  • Developer: Ilford MULTIGRADE, or BROMOPHEN, or PQ UNIVERSAL
  • Stop Bath : ILFOSTOP, or ILFOSTOP PRO
  • Fixer: ILFORE RAPID FIXER, or HYPAM
  • For ILFORD processors: ILFORD 2000RT developer/replenisher & fixer/replenisher are recommended
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty