Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iridium, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91015-IR 91014-IR 91007-IR 91020-IR 91009-IR 91016-IR 91013-IR 91010-IR 91006-IR 91018-IR 91008-IR 91019-IR
102099-556EA 312.85 USD InStock
102099-556 102099-436 102099-766 102099-524 102099-466 102099-586 102099-496 102099-706 102099-406 102099-736 102099-616 102099-646
Iridium, sputtering target sputter coater cathodes
Iridium
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.


Formula: Ir
MW: 192.22 g/mol
MDL Number: MFCD00011062
CAS Number: 7439-88-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty