Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences
Synonyms: Platinium

91015-PT 91006-PT 91018-PT 91016-PT 91013-PT 91009-PT 91017-PT 91007-PT 91019-PT 91020-PT 91012-PT 91008-PT 91014-PT
102099-446EA 993.43 USD InStock
102099-446 102099-776 102099-656 102099-686 102099-566 102099-596 102099-542 102099-476 102099-716 102099-626 102099-506 102099-416 102099-746
Platinum, sputtering target sputter coater cathodes
Platinum
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.


Formula: Pt
MW: 195.08 g/mol
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty