91014-CU
91013-CU
91018-CU
91007-CU
91019-CU
91015-CU
91017-CU
91010-CU
91016-CU
91009-CU
91008-CU
91020-CU
91006-CU
102099-732EA
417.53
USD
InStock
102099-732
102099-402
102099-612
102099-642
102099-552
102099-432
102099-520
102099-762
102099-462
102099-672
102099-702
102099-582
102099-492
Copper, sputtering target sputter coater cathodes
Copper
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.
EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.