Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91017-IR 91007-FE 91019-FE 91008-FE 91017-FE 91006-FE 91020-FE 91010-FE 91009-FE 91014-FE 91013-FE 91016-FE 91015-FE 91018-FE
102099-644EA 283.62 USD InStock
102099-644 102099-554 102099-676 102099-434 102099-764 102099-522 102099-464 102099-674 102099-584 102099-704 102099-734 102099-404 102099-614 102099-494
Iron, sputtering target sputter coater cathodes
Iron
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.


Formula: Fe
MW: 55.85 g/mol
MDL Number: MFCD00010999
CAS Number: 7439-89-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty