Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium, annular sputtering target, Diameter 87×57 mm (OD×ID)

Supplier: Electron Microscopy Sciences

91031-TI
102099-840EA 701.15 USD InStock
102099-840
Titanium, annular sputtering target, Diameter 87×57 mm (OD×ID)
Titanium
Metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


Formula: Ti
MW: 47.88 g/mol
MDL Number: MFCD00011264
CAS Number: 7440-32-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty