Would you like to log in again?
Gold Palladium, annular sputtering target, Diameter 59×57 mm (OD×ID)
Supplier: Electron Microscopy Sciences
91032-AP
102099-846EA 1252.62 USD InStock
102099-846
Gold Palladium, annular sputtering target, Diameter 59×57 mm (OD×ID)
Gold Palladium
Metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty