Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Carbon Film on Gilder Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF200F2-CU-50 CF200F2-NI-50 CF200F1-CU-ET CF200F2-AU-TH CF200F1-AU-25 CF200F2-NI-TH CF200F1-NI-50 CF200F1-CU-TH CF200F2-AU-25 CF200F2-CU-25 CF200F2-CU-UL CF200F2-CU-ET CF200F2-AU-ET CF200F1-AU-TH CF200F1-NI-ET CF200F1-AU-ET CF200F1-CU-25 CF200F1-NI-25 CF200F2-NI-ET CF200F2-AU-50 CF200F1-CU-50 CF200F1-CU-UL CF200F1-AU-UL CF200F1-NI-TH CF200F2-NI-25 CF200F2-CU-TH CF200F1-NI-UL CF200F2-NI-UL CF200F2-AU-UL CF200F1-AU-50
102099-996BX 241.59 USD InStock
102099-996 103303-622 102099-994 103303-666 102099-992 103303-668 103303-624 102099-990 103303-580 102099-998 102100-000 103304-386 103306-092 102100-002 102100-004 103306-058 103303-946 103303-948 103304-388 102099-986 103303-578 102099-984 102099-982 103305-766 103306-090 102099-988 103306-060 103304-494 103304-492 103305-768
Carbon Film on Gilder Finder Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select copper, nickel, or gold grids
Optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty