Would you like to log in again?
NOVA-TECH 10 Cobalt Blue, Sterile, Poly-Cellulose Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
NT10-1212CB NT10-99CB
10210-946BG 87.26 USD InStock
10210-946 10210-956
NOVA-TECH 10 Cobalt Blue, Sterile, Poly-Cellulose Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
NOVA-TECH 10 wipes are made of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform, super-tough fabric. These wipes are converted in a class 100 cleanroom.
  • Validated to SAL (Sterility Assurance Level) 10-6 in accordance with ANSI/AAMI/ISO 11137:2006 guidelines
  • Cobalt-60 gamma irradiated
  • ISO Class 5
  • Meets and exceeds USP 797 and USP 800 requirements for critical contamination control
  • Each packaged identified with lot number for traceability
  • Each case includes Certificate of Sterility
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty