A thin film of pure carbon is deposited on the shiny side of the grid.
- Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.