Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Carbon Film on Single Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFGA100-NI-TH CFGA150-NI-UL CFGA600-CU-TH CFGA300-NI-UL CFGA75-NI-UL CFGA800-CU-UL CFGA75-NI-25 CFGA1000-NI-50 CFGA600-NI-UL CFGA150-CU-UL CFGA400-CU-UL CFGA300-NI-TH CFGA600-NI-50 CFGA150-NI-50 CFGA100-CU-UL CFGA800-NI-TH CFGA400-NI-ET CFGA1000-CU-50 CFGA800-NI-50 CFGA100-NI-50 CFGA200-NI-25 CFGA200-NI-ET CFGA300-NI-ET CFGA800-CU-ET CFGA600-CU-50 CFGA300-NI-50 CFGA100-NI-UL CFGA150-CU-TH CFGA800-NI-ET CFGA1500-NI-25 CFGA200-NI-50 CFGA75-NI-ET CFGA200-NI-UL CFGA300-CU-TH CFGA200-CU-50 CFGA1000-CU-TH CFGA1500-CU-50 CFGA1000-NI-ET CFGA1000-CU-UL CFGA600-NI-ET CFGA300-CU-25 CFGA200-NI-TH CFGA200-CU-25 CFGA75-NI-50 CFGA400-CU-25 CFGA800-CU-50 CFGA300-CU-ET CFGA1500-CU-ET CFGA200-CU-UL CFGA300-CU-UL CFGA75-CU-25 CFGA400-NI-TH CFGA400-NI-UL CFGA1000-CU-ET CFGA1500-NI-ET CFGA150-CU-25 CFGA75-CU-ET CFGA800-NI-UL CFGA600-CU-ET CFGA800-NI-25 CFGA100-CU-TH CFGA150-CU-ET CFGA1000-NI-TH CFGA200-CU-ET CFGA400-CU-TH CFGA100-NI-ET CFGA1500-CU-25 CFGA150-CU-50 CFGA75-NI-TH CFGA800-CU-TH CFGA1000-CU-25 CFGA75-CU-50 CFGA100-CU-25 CFGA1000-NI-25 CFGA1000-NI-UL CFGA400-CU-50 CFGA300-CU-50 CFGA150-NI-25 CFGA100-CU-50 CFGA400-NI-25 CFGA100-CU-ET CFGA800-CU-25 CFGA150-NI-ET CFGA600-NI-TH CFGA100-NI-25 CFGA75-CU-UL CFGA1500-NI-UL CFGA1500-CU-TH CFGA400-NI-50 CFGA1500-NI-TH CFGA600-NI-25 CFGA1500-NI-50 CFGA200-CU-TH CFGA600-CU-UL CFGA400-CU-ET CFGA600-CU-25 CFGA75-CU-TH CFGA300-NI-25 CFGA150-NI-TH CFGA1500-CU-UL
103304-928PK 202.95 USD InStock
103304-928 103304-926 103304-408 103304-924 103304-406 102100-174 103304-484 102100-200 103304-482 102100-202 103304-480 102100-204 102100-206 103304-922 103304-404 103304-800 103304-920 102100-208 103304-402 103304-488 103304-400 103304-486 102100-180 102100-182 102100-184 102100-176 102100-178 102100-210 102100-212 102100-214 102100-216 102100-218 103304-478 102100-190 102100-192 102100-194 102100-196 102100-220 102100-186 102100-222 102100-188 102100-224 102100-226 102100-228 103304-860 103304-858 102100-240 102100-198 102100-230 102100-232 102100-234 102100-236 103304-856 102100-238 103304-854 103304-852 103304-850 103304-848 103304-846 102100-250 102100-242 102100-244 102100-246 102100-248 103304-844 103304-842 103304-034 103304-792 102100-252 103304-396 103304-032 103304-790 103304-394 103304-030 103304-798 103304-038 103304-796 103304-036 103304-794 103304-398 103304-782 103304-022 103304-780 103304-020 103304-788 103304-028 103304-786 103304-026 103304-784 103304-024 103304-938 103304-936 103304-934 103304-490 103304-778 103304-932 103304-776 103304-930 103304-412 103304-410
Carbon Film on Single Hole Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 – 6 nm), thick (15 – 20 nm), or extra thick (25 – 50 nm)
  • Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes
Optically checked followed by batch testing in the electron microscope.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty