A thin film of pure carbon is deposited on the shiny side of the grid.
- Carbon coatings thickness of standard (10 nm), ultra-thin (5 – 6 nm), thick (15 – 20 nm), or extra thick (25 – 50 nm)
- Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes
Optically checked followed by batch testing in the electron microscope.
Packaging: Packed in grid storage box.