A thin film of pure carbon is deposited on the shiny side of the grid.
- Select copper, nickel, or gold grids in 200, 300, 4000, or 1000 mesh
- Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 - 6 nm), thick (15 - 20 nm), or extra thick (25 - 50 nm).
Packaging: Packed in grid storage box.