Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Formvar Films on London Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FFLF135-NI-ET FFLF200-AU-25 FFLF135-AU-ET FFLF135-CU-TH FFLF200-CU-50 FFLF200-CU-ET FFLF135-AU-25 FFLF200-NI-UL FFLF135-CU-ET FFLF400-NI-TH FFLF135-CU-UL FFLF135-AU-UL FFLF400-CU-25 FFLF200-CU-TH FFLF135-NI-25 FFLF400-CU-50 FFLF200-CU-UL FFLF400-CU-UL FFLF135-NI-UL FFLF400-CU-ET FFLF400-AU-25 FFLF200-CU-25 FFLF135-CU-25 FFLF200-NI-25 FFLF200-NI-TH FFLF400-NI-25 FFLF400-NI-50 FFLF135-AU-50 FFLF135-CU-50 FFLF200-NI-50 FFLF400-NI-ET FFLF400-NI-UL FFLF135-NI-50 FFLF200-NI-ET FFLF135-NI-TH FFLF135-AU-TH FFLF400-CU-TH
103303-846PK 166.23 USD InStock
103303-846 103305-710 103305-932 103305-930 103305-612 102100-960 103304-084 102100-962 103304-082 102100-940 103305-934 103303-848 102100-964 102100-942 102100-966 102100-944 102100-946 102100-968 102100-948 103305-042 103303-850 103305-044 103305-040 103304-080 103305-104 103305-106 103304-374 102100-950 103305-108 102100-952 102100-954 102100-956 102100-958 103306-006 103304-378 102100-938 103304-376
Formvar Films on London Finder Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Thin film of pure formvar resin coats the shiny side of a London finder grids.
  • Select from copper, nickel, and gold grids
  • Optically checked, followed by batch testing in the EM
Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty