Would you like to log in again?
Formvar/Silicone Monoxide Support Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FSF300-NI FSF400-CU FSF300-CU FSF200-CU FSF400-NI FSF200-NI
102101-074BX 352.25 USD InStock
102101-074 102101-072 102101-070 102101-068 102101-066 102101-076
Formvar/Silicone Monoxide Support Films, Electron Microscopy Sciences
Graticules Reticles and Grids
A formvar film stabilized with a thin film of Silicone monoxide.
  • Desirable support film
  • Low background contrast
  • Stable under the electron beam
  • More hydrophilic than carbon film
All of coated grids are optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.
Caution: Note: All of our film is laid on the shiny side of the grid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty