Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Gilder Grids Standard Square Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G2010-MO G200-MO G100-MO G1000-MO
102101-140PK 121.96 USD InStock
102101-140 102101-080 102101-132 102101-086
Gilder Grids Standard Square Mesh, Electron Microscopy Sciences
Graticules Reticles and Grids
The products are manufactured using a process known as chemical ʽmilling’ (etching) instead of the more familiar technique of ʽelectroforming’ (deposition) that is used in the manufacture of copper, nickel and gold products. Molybdenum is principally used.
  • High temperature
  • Hardness
  • High expansion coefficient
  • Corrosion resistance
The material which is used has a purity of 99.9%.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty