Would you like to log in again?
Parallel Bars Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G300P-AU G75PB-CU G200P-CU G100PB-NI G300P-NI G300PB-NI G200PB-CU G200PB-NI G75PB-AU G75P-CU G100P-CU G400PB-AU G300PB-CU G400P-NI G100PB-AU G50P-NI G100P-AU G75PB-NI G300PB-AU G50P-CU G300P-CU G100P-NI G100PB-CU G50PB-NI G75P-AU G400PB-NI G75P-NI G150P-AU G400PB-CU G50PB-AU G50PB-CU G200P-NI G200P-AU G50P-AU G400P-AU G150PB-NI G150PB-AU G150P-CU G150P-NI G150PB-CU G400P-CU G200PB-AU
100495-686EA 61.3 USD InStock
100495-686 102101-192 102101-190 102101-090 100495-626 100495-628 100495-688 102101-108 102101-106 102101-104 102101-168 102101-102 100495-690 100503-004 102101-188 102101-100 102101-166 102101-088 102101-220 100503-002 102101-164 102101-186 102101-162 102101-184 100495-630 102101-182 100495-632 100502-994 100502-992 102101-218 102101-216 102101-214 100501-524 102101-136 102101-212 100501-522 100501-544 102101-134 102101-210 100501-542 102101-110 100501-540
Parallel Bars Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt or shiny side. Each grid is individually inspected.
  • 3.05 mm diameter
  • 0.7 mil (18 µm) thickness
  • In Copper (Cu), Nickel (Ni), or Gold (Au)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty