Would you like to log in again?
Single Oval Slot Grids and Slot Grids with Support Bar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G102-CU G153-CU G2010-CP G2010-CU G60610-MO G153-NI G102-NI G215-AU G205-AU G215-NI G60610-NI G205-NI G205-CU G207-CU G2010-NI G153-AU G215-CU G207-NI G207-AU G60610-CU G60610-AU G2010-AU G102-AU
102101-096PK 75.6 USD InStock
102101-096 102101-150 102101-194 102101-094 102101-092 100495-670 100501-528 100495-668 102101-116 102101-138 102101-148 102101-114 102101-146 102101-200 102101-112 100501-530 102101-144 103302-640 102101-198 102101-154 102101-142 102101-152 102101-196
Single Oval Slot Grids and Slot Grids with Support Bar, Electron Microscopy Sciences
Graticules Reticles and Grids
Gilder grids are a reliable support specimen grid for transmission electron microscopy.
  • Copper, copper-palladium, nickel, molybdenum, and gold
  • Grids with two central support bars enable thinner films
  • Each grid is individually inspected
Single Slot TEM Grids with oval holes have a thickness of are 50 microns.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty