Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Gilder Standard Square Mesh Grids, Gilder Grids
Supplier: Electron Microscopy Sciences
G100-CU G200-CP G250-CU G200-CU G300-CU G75-NI G50-CU G200TT-CU G150-NI G200-NI G175-AU G50-NI G200TT-NI G400-CP G200T-NI G200TH-CU G75-CU G200T-CU G200TH-NI G50-AU G175-NI G150-CU G300-AU G300-CP G100-NI G400-AU G75-AU G250-NI G100-AU G150-AU G150-CP G100-CP G250-AU G175-CU G400-NI G400-CU G200-AU G300-NI
100495-662EA 46.89 USD InStock
100495-662 100495-684 102101-172 100503-006 100495-664 102101-170 100501-514 100495-660 100495-666 100495-624 100502-986 100501-534 100501-512 100503-000 102101-202 100501-510 100501-532 102101-122 102101-120 100495-674 102101-160 102101-082 100495-676 102101-180 100495-692 100495-694 100501-526 100495-672 100495-616 100495-618 100502-996 100502-990 100495-634 100495-636 102101-118 102101-158 102101-156 102101-098
Gilder Standard Square Mesh Grids, Gilder Grids
Graticules Reticles and Grids
In Copper (Cu), Nickel (Ni), Gold (Au), Copper/Palladium (Cu/Pd=CP), Molybdenum (Mo).
  • Measures 3.05 mm in diameter
  • 0.7 mil (18 µm) thickness
G200TH is a thickened version of the standard, with an asymmetric centre. A mark on the rim allows for precise orientation of the grids.
G200TT is a combination of thin and thick bar grids, with a mark on the rim for orientation.
G200T is designed for ease of handling and is easily removed if necessary. To remove the handle, just bend on a 90 degree angle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty