Would you like to log in again?
Gilder Standard Hexagonal Mesh Grids, Gilder Grids
Supplier: Electron Microscopy Sciences
G100H-CU G200H-AU G300H-AU G200H-CU G50H-AU G75H-CU G75H-NI G50H-CU G300H-NI G100H-NI G200H-NI G300H-CU G400H-AU G400H-NI G50H-NI G400H-CU G75H-AU G100H-AU
102101-084PK 88.34 USD InStock
102101-084 100495-620 100501-518 100501-516 100501-538 100495-680 100501-536 100495-682 100495-622 100495-678 102101-208 102101-206 100502-998 100502-988 102101-204 102101-178 102101-176 102101-174
Gilder Standard Hexagonal Mesh Grids, Gilder Grids
Graticules, Reticles and Grids
A reliable support specimen grid source.
  • Measures 3.05 mm in diameter
  • Material: Copper (Cu), Nickel (Ni), Gold (Au)
Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty