Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Single Slot Grids (Aperture Grids), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
GA100-CU GA400-NI GA500-AU GA200-NI GA400-AU GA2000-AU GA2000-CU GA50-NI GA1500-NI GA1000-NI GA1000-MO GA600-AU GA500-CU GA800-NI GA500-NI GA1500-CU GA2000-NI GA150-CU GA1000-CU GA1000-AU GA50-AU GA50-CU GA600-CU GA400-CU GA300-AU GA75-NI GA800-AU GA300-NI GA75-CU GA100-NI GA200-CU GA600-NI GA150-NI GA150-AU GA300-CU GA200-AU GA1500-AU GA100-AU GA800-CU GA75-AU
102101-250PK 54.54 USD InStock
102101-250 100495-720 100495-704 100495-706 100495-700 100495-702 100495-708 102101-228 102101-248 102101-226 102101-246 102101-224 102101-244 103302-194 103302-150 102101-266 102101-222 102101-264 102101-242 100501-550 102101-240 102101-262 102101-260 100495-696 100495-698 100501-548 100495-714 100495-716 100495-710 100495-712 100495-718 102101-238 102101-258 102101-236 102101-256 102101-234 102101-232 102101-254 102101-252 102101-230
Single Slot Grids (Aperture Grids), Electron Microscopy Sciences
Graticules Reticles and Grids
Gilder aperture grids with a single slot.
  • Copper, nickel, molybdenum, and gold grids
  • Holes sizes of 50, 75, 100, 150, 200, 300, 400, 600, 800, 1000, 1500, and 2000 microns
  • 50 micron thickness
Copper, nickel, molybdenum, and gold grids
Holes sizes of 50, 75, 100, 150, 200, 300, 400, 600, 800, 1000, 1500, and 2000 microns
50 micron thickness
Single Slot aperture TEM grids have a thickness of are 50 microns. Each grid is individually inspected.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty