Lacy carbon film provides support without interfering with transmission electron microscopy observation.
- Copper, nickel and gold grids in 200, 300, and 400 mesh grids
- Optically checked followed by batch testing in the EM
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.