Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicon(II) oxide on Cu grid, support films 15 – 30 nm for electron microscopy

Supplier: Electron Microscopy Sciences
Synonyms: Silicium (II) oxideSilicon monoxideSilicon oxide

SF400-CU SF200-CU SF300-CU
102101-536PK 364.98 USD InStock
102101-536 102101-532 102101-540
Silicon(II) oxide on Cu grid, support films 15 – 30 nm for electron microscopy
Silicon (II) oxide
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM.

A thin film of pure silicone Monoxide (15 – 30 nm) is deposited directly on top of the grid. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. All of our film is laid on the shiny side of the grid.


Formula: SiO
MW: 44.08 g/mol
MDL Number: MFCD00151536
CAS Number: 10097-28-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty