Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
QuantomiX Wet SEM Capsules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
RT-58 RT-56 QX-302-6 SK-102-302 MA-4 SP-202-48 IB-74 IB-64 SK-202-24 SK-302-3 SK-102-24 SP-202-24 PI-24 MP-10-2 MP-12-2 MM-25 SP-102-24 SK-302-12 SK-202-48 QX-102-24
102101-518EA 385.25 USD InStock
102101-518 102101-516 102101-504 102101-514 102101-546 102101-458 102101-568 102101-556 102101-544 102101-478 102101-554 102101-520 102101-486 102101-552 102101-364 102101-572 102101-550 102101-484 102101-362 102101-570
QuantomiX Wet SEM Capsules, Electron Microscopy Sciences
Culture Dishes Cell Culture Dishes
The QX-102 Capsules are used for imaging various wet materials and biological samples, such as liquid samples (foods, cosmetics, oils, paints, etc.), particles in solutions, adherent and non-adherent human and animal cells, and microorganisms.
  • No coating or embedding of the sample are required
  • Enables electron microscopy imaging with easy sample preparation comparable to light microscopy
The samples can be visualized either directly or by following appropriate contrast enhancement staining or labeling procedures. The capsule serves as a cell culture dish, specifically designed for the SEM. The QX-102 capsules are supplied sterile for single use.
Packaging: Packed in boxes of 24.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty