Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DUSP23 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40452
102121-858EA 564.74 USD InStock
102121-858
Anti-DUSP23 Rabbit Polyclonal Antibody
Antibodies
The DUSP23 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DUSP23. This antibody reacts with human, rat, bat, bovine, equine, monkey. The DUSP23 Antibody has been validated for the following applications: ELISA, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: DUSP23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bat, Bovine, Equine, Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty