Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB110-60935B
102124-274EA 614.15 USD InStock
102124-274
Anti-FTO Rabbit Polyclonal Antibody (Biotin)
Antibodies
The FTO Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to FTO. This antibody reacts with human, mouse, primate. The FTO Antibody [Biotin] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: FTO
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primate

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty