Would you like to log in again?
Pepsin
Supplier: Novus Biologicals
NB900-66724
102136-948EA 352.43 USD InStock
102136-948
Pepsin
Enzymes
Stablized Pepsin solution for antigen retrieval
  • No Preservative
Dilutions: Immunohistochemistry 1:10-1:500
Application Notes: 1. Deparaffinize the tissue sections and bring them to buffer. 2. Remove excess buffer from the glass without letting the tissue dry. 3. Depending on the size of the tissue section, add 1 or 2 drops of pepsin solution. 4. Incubate the slide for 5 to 10 minutes at 37C. 5. Wash slides with buffer and proceed for the immunostaining.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty