Would you like to log in again?
Fluoromount Mounting Media, Novus Biologicals
Supplier: Novus Biologicals
Fluoromount™
NB900-66726
102136-950EA 281.66 USD InStock
102136-950
Fluoromount Mounting Media, Novus Biologicals
Stains Microscopy and staining reagents
Aqueous mounting medium for fluorescent staining
  • Preservative: Sodium Azide
Fluoromount is an aqueous based mounting medium for cover slipping tissues stained with various fluorescent dyes. This product is compatible with FITC, phycoerythrin, phycocyanin and allophycocyanin. However, it can be used with other fluorescent markers such as Texas Red and rhodamine.
Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty