Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant Phosphoserine phosphatase (from E. coli)

Supplier: Novus Biologicals
NBC1-18384
102137-126EA 578.89 USD InStock
102137-126
Human Recombinant Phosphoserine phosphatase (from E. coli)
Enzymes
The Recombinant Human Phosphoserine phosphatase Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Phosphoserine phosphatase Protein has been validated for the following applications: SDS-Page.

  • Entrez Gene ID: 5723

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty