Would you like to log in again?
SD OLLAS/DYKDDDDK Epitope Tag Kit, Novus Biologicals (NBK1-19035)
Supplier: Novus Biologicals
NBK1-19035
102137-700EA 491.08 USD InStock
102137-700
SD OLLAS/DYKDDDDK Epitope Tag Kit, Novus Biologicals (NBK1-19035)
Assays Immunological Assays
SD OLLAS/DYKDDDDK Epitope Tag Kit
Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Non-species specific.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty