Would you like to log in again?
CritiCore® ESD Stripe Fluid-Resistant Barrier Coats
Supplier: CritiCore Protective Wear
10215-536 5059BARENVYM 10215-527 10215-538 5059BARENVY4XL 5059BARENVYS 10215-542 10215-534 10215-540 5059BARENVYL 10215-524 5059BARENVY2XL 5059BARENVYXXS 10215-530 5059BARENVY5XL 5059BARENVY3XL 5059BARENVYXS 5059BARENVY6XL 5059BARENVYXL 10215-528 10215-532
76422-832CS 1201.65 USD InStock
76422-832 10215-529 76422-842 10215-527 10215-538 76422-830 76422-840 10215-536 10215-525 10215-534 10215-532 76422-828 76422-838 10215-542 76422-826 10215-531 76422-836 10215-540 76422-846 76422-834 76422-844
CritiCore® ESD Stripe Fluid-Resistant Barrier Coats
Workwear
ESD barrier lab coats are made with a fluid-resistant material consisting of 99% polyester and a 1% carbon stripe for electrostatic dissipation (ESD). These barrier coats are ideal for use in medical, healthcare, veterinary, and food processing applications.
  • One breast pocket
  • Two oversized waist pockets
  • Cuffed sleeves
  • Six snap closure
  • Side seam openings for easy access to pants pockets
These coats are extremely durable and will hold up to repeated industrial launderings.
Ordering information: Coats in sizes X-small to XX-large are stocked. For sizes 3X-large to 6X-large, extended lead time may be required. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty