Would you like to log in again?
Autosampler Vials, without Caps, DWK Life Sciences
Supplier: DWK Life Sciences
511232SNAW 511232SAW 331232SNW 331232CLW 511232SA 331232S 331545S 511545SA 331232SW 511232CLAW
10061-480CS 969.75 USD InStock
10061-480 10061-472 10216-864 10061-470 50880-006 10061-482 10061-474 10216-862 10061-478 10061-468
Autosampler Vials, without Caps, DWK Life Sciences
Vials
These clear glass autosampler vials are manufactured from 33 expansion, low extractable borosilicate glass.
  • Snap cap opening with marking spot
  • Graduated
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Closures not included. Available in a variety of standard sizes and finishes. Vials with large opening and/or marking spot are also available.
Packaging: Packed 100 per tray.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty