Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant Cysteine Dioxygenase Type 1 (from E. coli)

Supplier: Novus Biologicals
NBP1-30241
102163-236EA 423.2 USD InStock
102163-236
Human Recombinant Cysteine Dioxygenase Type 1 (from E. coli)
Enzymes
The Recombinant Human Cysteine Dioxygenase Type 1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Cysteine Dioxygenase Type 1 Protein has been validated for the following applications: SDS-Page.

  • Entrez Gene ID: 1036

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty