Would you like to log in again?
Sampler, MiniSampler, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6205-0090 5305-0010 5305-0011 5305-1020 5305-0110 6205-0180 5305-0209 5305-0100
76047-098EA 129.05 USD InStock
76047-098 10217-038 10217-032 10217-040 10217-036 10217-044 76047-100 10217-034
Sampler, MiniSampler, Bürkle
Samplers Fluid Samplers
The single-use, flexible, suction hose can be quickly replaced, thus ensuring that the samples are never contaminated
If necessary, a new hose can be used for every single sample. Due to its small diameter (8mm) and flexibility, the hose can reach sampling areas which are otherwise inaccessible. Maximum suction lift is five meters, depending on viscosity.
Accessories information: Accessory sample containers include a screw lid, and are ideal for sampling and transport. Made of PFA for super pure collection of samples, containers are autoclavable.
Delivery information: Supplied complete with one vacuum pump, 10 PE bottles, PE suction hose, one hose cutter, one hose weight made of stainless steel, and 20 seal-it bottle seals.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty